Product Details

SNP ID
rs201650288
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4143917 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGCGGCGGCCCAGGGGACCCGGG[C/T]CTGTACTTGGCGTTGCTCGGCCGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYB5D2 PubMed Links

Gene Details

Gene
CYB5D2
Gene Name
cytochrome b5 domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001254755.1 444 Intron NP_001241684.1
NM_001254756.1 444 Intron NP_001241685.1
NM_144611.3 444 Silent Mutation GGC,GGT G54G NP_653212.1
XM_006721447.2 444 Intron XP_006721510.2
XM_011523649.2 444 Intron XP_011521951.1
XM_017024180.1 444 Intron XP_016879669.1
XM_017024181.1 444 Intron XP_016879670.1
XM_017024182.1 444 Intron XP_016879671.1
XM_017024183.1 444 Silent Mutation GGC,GGT G54G XP_016879672.1
Gene
ZZEF1
Gene Name
zinc finger ZZ-type and EF-hand domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015113.3 444 Intron NP_055928.3
XM_005256561.2 444 Intron XP_005256618.1
XM_011523759.2 444 Intron XP_011522061.1
XM_017024382.1 444 Intron XP_016879871.1
XM_017024383.1 444 Intron XP_016879872.1
XM_017024384.1 444 Intron XP_016879873.1

View Full Product Details