Product Details

SNP ID
rs201308324
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:47832172 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCTTATCACTAAGCGGAACTTGA[C/G]TTTCCCTGAAGATGGGGAACTGTCA
Phenotype
MIM: 611825 MIM: 614966
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LRRC46 PubMed Links

Gene Details

Gene
LRRC46
Gene Name
leucine rich repeat containing 46
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033413.3 446 Missense Mutation ACT,AGT T28S NP_219481.1
XM_005257775.4 446 Intron XP_005257832.1
Gene
MRPL10
Gene Name
mitochondrial ribosomal protein L10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145255.3 446 Intron NP_660298.2
NM_148887.2 446 Intron NP_683685.1
Gene
SCRN2
Gene Name
secernin 2
There are no transcripts associated with this gene.

View Full Product Details