Product Details

SNP ID
rs202203881
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:5106240 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATAAATGAAGGTTTTACCACATT[C/T]ACTACATTCATGGTCTTTCTTCCCT
Phenotype
MIM: 194480 MIM: 616463
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928000 PubMed Links

Gene Details

Gene
LOC101928000
Gene Name
uncharacterized LOC101928000
There are no transcripts associated with this gene.

Gene
ZFP3
Gene Name
ZFP3 zinc finger protein
There are no transcripts associated with this gene.

Gene
ZNF232
Gene Name
zinc finger protein 232
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320952.1 1037 Missense Mutation AAA,GAA K298E NP_001307881.1
NM_001320953.1 1037 Missense Mutation AAA,GAA K280E NP_001307882.1
NM_001320954.1 1037 Missense Mutation AAA,GAA K280E NP_001307883.1
NM_001320955.1 1037 Missense Mutation AAA,GAA K279E NP_001307884.1
NM_014519.3 1037 Missense Mutation AAA,GAA K307E NP_055334.2
XM_011524006.2 1037 Missense Mutation AAA,GAA K259E XP_011522308.1
XM_017025021.1 1037 Missense Mutation AAA,GAA K307E XP_016880510.1
XM_017025022.1 1037 Missense Mutation AAA,GAA K231E XP_016880511.1
XM_017025023.1 1037 Missense Mutation AAA,GAA K222E XP_016880512.1

View Full Product Details