Product Details

SNP ID
rs201004949
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:18184608 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGAGAAGGGCCTGTACAACGAGC[A/G]CACGGTGGACCGGGCCCCACTGCGC
Phenotype
MIM: 613303 MIM: 602666
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALKBH5 PubMed Links

Gene Details

Gene
ALKBH5
Gene Name
alkB homolog 5, RNA demethylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017758.3 1056 Missense Mutation CAC,CGC H122R NP_060228.3
Gene
MYO15A
Gene Name
myosin XVA
There are no transcripts associated with this gene.

View Full Product Details