Product Details
- SNP ID
-
rs201448609
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:45440186 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCTGTTGGAGGTCAGCAACACTGA[A/G]CCTATGCGGAGATTCCAAGAGATAA
- Phenotype
-
MIM: 610591
MIM: 611466
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ARHGAP27
PubMed Links
Gene Details
- Gene
- ARHGAP27
- Gene Name
- Rho GTPase activating protein 27
There are no transcripts associated with this gene.
- Gene
- PLEKHM1
- Gene Name
- pleckstrin homology and RUN domain containing M1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014798.2 |
2751 |
Missense Mutation |
CTC,TTC |
L960F |
NP_055613.1 |
XM_006722201.3 |
2751 |
Missense Mutation |
CTC,TTC |
L830F |
XP_006722264.1 |
XM_011525523.2 |
2751 |
Intron |
|
|
XP_011523825.1 |
XM_011525524.1 |
2751 |
Missense Mutation |
CTC,TTC |
L960F |
XP_011523826.1 |
XM_011525525.1 |
2751 |
Missense Mutation |
CTC,TTC |
L909F |
XP_011523827.1 |
XM_011525526.1 |
2751 |
Missense Mutation |
CTC,TTC |
L893F |
XP_011523828.1 |
XM_011525528.2 |
2751 |
Missense Mutation |
CTC,TTC |
L439F |
XP_011523830.1 |
XM_017025451.1 |
2751 |
Missense Mutation |
CTC,TTC |
L960F |
XP_016880940.1 |
XM_017025452.1 |
2751 |
Missense Mutation |
CTC,TTC |
L871F |
XP_016880941.1 |
XM_017025453.1 |
2751 |
Missense Mutation |
CTC,TTC |
L830F |
XP_016880942.1 |
XM_017025454.1 |
2751 |
Missense Mutation |
CTC,TTC |
L435F |
XP_016880943.1 |
View Full Product Details