Product Details

SNP ID
rs201653455
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42955224 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCACATGATCCTCTGCTCCACA[C/T]CTGAAAGAGAAAGGTCAGAGGAGAC
Phenotype
MIM: 613212
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AARSD1 PubMed Links

Gene Details

Gene
AARSD1
Gene Name
alanyl-tRNA synthetase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261434.1 1663 Silent Mutation NP_001248363.1
Gene
PTGES3L-AARSD1
Gene Name
PTGES3L-AARSD1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136042.2 1663 Silent Mutation NP_001129514.2
NM_025267.3 1663 Silent Mutation NP_079543.1

View Full Product Details