Product Details

SNP ID
rs200731218
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:49578779 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGGGGAGGCTTGGGGCATTCTT[A/G]GGCAGCCCCCCAACCGCCCGAACCA
Phenotype
MIM: 604636
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100288866 PubMed Links

Gene Details

Gene
LOC100288866
Gene Name
uncharacterized LOC100288866
There are no transcripts associated with this gene.

Gene
NXPH3
Gene Name
neurexophilin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007225.2 522 Missense Mutation AGG,GGG R80G NP_009156.2

View Full Product Details