Product Details

SNP ID
rs201830704
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:43013320 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACACGATCAACATGGAGGAGTGC[C/G]GGCTGCGGGTGCAGGTCCAGCCCTT
Phenotype
MIM: 600735 MIM: 604631
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IFI35 PubMed Links

Gene Details

Gene
IFI35
Gene Name
interferon induced protein 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005533.4 530 Missense Mutation CGG,GGG R108G NP_005524.2
XM_005257302.4 530 Missense Mutation CGG,GGG R108G XP_005257359.1
XM_017024584.1 530 Missense Mutation CGG,GGG R88G XP_016880073.1
XM_017024585.1 530 Missense Mutation CGG,GGG R88G XP_016880074.1
Gene
VAT1
Gene Name
vesicle amine transport 1
There are no transcripts associated with this gene.

View Full Product Details