Product Details

SNP ID
rs201556743
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44926100 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGGTCAGGACACCTTGGTGACGC[C/T]GTTCCCTGCTGAGAGTGCTCTCCCC
Phenotype
MIM: 614570
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIF18B PubMed Links

Gene Details

Gene
KIF18B
Gene Name
kinesin family member 18B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001264573.1 2708 UTR 3 NP_001251503.1
NM_001265577.1 2708 Missense Mutation AGC,GGC S847G NP_001252506.1
XM_011524385.2 2708 Missense Mutation AGC,GGC S868G XP_011522687.1
XM_011524386.2 2708 Missense Mutation AGC,GGC S859G XP_011522688.1
XM_011524387.2 2708 Missense Mutation AGC,GGC S859G XP_011522689.1
XM_011524388.2 2708 Missense Mutation AGC,GGC S856G XP_011522690.1
XM_011524389.2 2708 UTR 3 XP_011522691.1
XM_011524390.2 2708 Missense Mutation AGC,GGC S833G XP_011522692.1
XM_011524391.2 2708 Intron XP_011522693.1
Gene
MIR6783
Gene Name
microRNA 6783
There are no transcripts associated with this gene.

View Full Product Details