Product Details

SNP ID
rs200875152
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4771998 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCTCGTCTGCATTGTGGCCAAC[A/G]CCCTCCTGCTGGTACCTAATGGGGA
Phenotype
MIM: 604657
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TM4SF5 PubMed Links

Gene Details

Gene
TM4SF5
Gene Name
transmembrane 4 L six family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003963.2 107 Missense Mutation ACC,GCC T26A NP_003954.2

View Full Product Details