Product Details

SNP ID
rs201728723
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81905393 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCATGACCCACCTGGTATGGGT[C/T]GGAGCCATCCCTGTCAGGGATAATT
Phenotype
MIM: 614534 MIM: 607996 MIM: 602679 MIM: 606212
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANAPC11 PubMed Links

Gene Details

Gene
ANAPC11
Gene Name
anaphase promoting complex subunit 11
There are no transcripts associated with this gene.

Gene
NPB
Gene Name
neuropeptide B
There are no transcripts associated with this gene.

Gene
PCYT2
Gene Name
phosphate cytidylyltransferase 2, ethanolamine
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184917.2 1364 Missense Mutation AAC,GAC N338D NP_001171846.1
NM_001256433.2 1364 Missense Mutation AAC,GAC N266D NP_001243362.1
NM_001256434.2 1364 Missense Mutation AAC,GAC N279D NP_001243363.1
NM_001256435.2 1364 Missense Mutation AAC,GAC N242D NP_001243364.1
NM_001282203.1 1364 Missense Mutation AAC,GAC N242D NP_001269132.1
NM_001282204.1 1364 Missense Mutation AAC,GAC N288D NP_001269133.1
NM_002861.4 1364 Missense Mutation AAC,GAC N320D NP_002852.1
XM_005256386.3 1364 Missense Mutation AAC,GAC N306D XP_005256443.1
XM_005256387.3 1364 Missense Mutation AAC,GAC N306D XP_005256444.1
XM_006722287.3 1364 Intron XP_006722350.1
XM_017024910.1 1364 Intron XP_016880399.1
XM_017024911.1 1364 Missense Mutation AAC,GAC N288D XP_016880400.1
XM_017024912.1 1364 Missense Mutation AAC,GAC N151D XP_016880401.1
Gene
SIRT7
Gene Name
sirtuin 7
There are no transcripts associated with this gene.

View Full Product Details