Product Details
- SNP ID
-
rs202179083
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:74772263 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACGGCCTCAGTGCCAAGGCCCTTAC[C/T]CCTGCAGCTGGGCTCTAGGAGAGAC
- Phenotype
-
MIM: 604990
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NAT9
PubMed Links
Gene Details
- Gene
- NAT9
- Gene Name
- N-acetyltransferase 9 (putative)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001305077.1 |
443 |
Missense Mutation |
AGT,GGT |
S116G |
NP_001292006.1 |
NM_001305078.1 |
443 |
Missense Mutation |
AGT,GGT |
S117G |
NP_001292007.1 |
NM_001305079.1 |
443 |
Missense Mutation |
AGT,GGT |
S117G |
NP_001292008.1 |
NM_001305080.1 |
443 |
Missense Mutation |
AGT,GGT |
S117G |
NP_001292009.1 |
NM_001305081.1 |
443 |
Missense Mutation |
AGT,GGT |
S116G |
NP_001292010.1 |
NM_001305082.1 |
443 |
Missense Mutation |
AGT,GGT |
S122G |
NP_001292011.1 |
NM_001305083.1 |
443 |
Missense Mutation |
AGT,GGT |
S116G |
NP_001292012.1 |
NM_001305084.1 |
443 |
Missense Mutation |
AGT,GGT |
S117G |
NP_001292013.1 |
NM_001305085.1 |
443 |
Missense Mutation |
AGT,GGT |
S116G |
NP_001292014.1 |
NM_001305086.1 |
443 |
Missense Mutation |
AGT,GGT |
S116G |
NP_001292015.1 |
NM_001305087.1 |
443 |
Missense Mutation |
AGT,GGT |
S92G |
NP_001292016.1 |
NM_001305088.1 |
443 |
Missense Mutation |
AGT,GGT |
S86G |
NP_001292017.1 |
NM_015654.4 |
443 |
Missense Mutation |
AGT,GGT |
S117G |
NP_056469.2 |
XM_005257212.4 |
443 |
Missense Mutation |
AGT,GGT |
S86G |
XP_005257269.1 |
XM_006721813.3 |
443 |
Missense Mutation |
AGT,GGT |
S86G |
XP_006721876.1 |
XM_011524607.2 |
443 |
Intron |
|
|
XP_011522909.1 |
XM_017024437.1 |
443 |
Missense Mutation |
AGT,GGT |
S92G |
XP_016879926.1 |
XM_017024438.1 |
443 |
Intron |
|
|
XP_016879927.1 |
XM_017024439.1 |
443 |
Missense Mutation |
AGT,GGT |
S116G |
XP_016879928.1 |
XM_017024440.1 |
443 |
Intron |
|
|
XP_016879929.1 |
XM_017024441.1 |
443 |
Missense Mutation |
AGT,GGT |
S86G |
XP_016879930.1 |
- Gene
- SLC9A3R1
- Gene Name
- SLC9A3 regulator 1
There are no transcripts associated with this gene.
- Gene
- TMEM104
- Gene Name
- transmembrane protein 104
There are no transcripts associated with this gene.
View Full Product Details