Product Details

SNP ID
rs200947207
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:73285732 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCTTGCCAGGGGAGGGGCCGCCA[C/T]GGCGTACGGGGGAGCCTGGGTGATG
Phenotype
MIM: 605468
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDC42EP4 PubMed Links

Gene Details

Gene
CDC42EP4
Gene Name
CDC42 effector protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012121.4 1308 Missense Mutation ATG,GTG M257V NP_036253.2
XM_005257182.2 1308 Missense Mutation ATG,GTG M257V XP_005257239.1

View Full Product Details