Product Details

SNP ID
rs201492595
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8161707 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCATCTGCACGGTCGTCCAGCTC[C/T]GACAGCTTCTGGTCTCGCTCCAGGA
Phenotype
MIM: 602260 MIM: 185881
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PER1 PubMed Links

Gene Details

Gene
PER1
Gene Name
period circadian clock 1
There are no transcripts associated with this gene.

Gene
VAMP2
Gene Name
vesicle associated membrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014232.2 609 Silent Mutation NP_055047.2
XM_005256775.4 609 Silent Mutation XP_005256832.1

View Full Product Details