Product Details

SNP ID
rs200741294
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1651304 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCGTCAGTGTACAGGAGCCTGG[C/T]GTGAAGCTGGGGGAGGAACGAGGAC
Phenotype
MIM: 607300 MIM: 607848 MIM: 607873
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRPF8 PubMed Links

Gene Details

Gene
PRPF8
Gene Name
pre-mRNA processing factor 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006445.3 6771 Silent Mutation ACC,ACT T2219T NP_006436.3
Gene
RILP
Gene Name
Rab interacting lysosomal protein
There are no transcripts associated with this gene.

Gene
SCARF1
Gene Name
scavenger receptor class F member 1
There are no transcripts associated with this gene.

View Full Product Details