Product Details

SNP ID
rs201085963
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7252569 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGGAGATGTTGGACTCGCTGTTG[A/G]CCTTGGGCGGCCTGGTGCTGCTTCG
Phenotype
MIM: 609131 MIM: 610684 MIM: 615019
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLDN7 PubMed Links

Gene Details

Gene
CLDN7
Gene Name
claudin 7
There are no transcripts associated with this gene.

Gene
CTDNEP1
Gene Name
CTD nuclear envelope phosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143775.1 281 Intron NP_001137247.1
NM_015343.4 281 Intron NP_056158.2
Gene
ELP5
Gene Name
elongator acetyltransferase complex subunit 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015362.4 281 Missense Mutation ACC,GCC T23A NP_056177.3
NM_203413.2 281 Missense Mutation ACC,GCC T23A NP_981958.1
NM_203414.2 281 Missense Mutation ACC,GCC T23A NP_981959.1
NM_203415.1 281 Missense Mutation ACC,GCC T23A NP_981960.1
XM_011523779.2 281 Missense Mutation ACC,GCC T23A XP_011522081.1

View Full Product Details