Product Details

SNP ID
rs200879867
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7194492 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGGTCTTGTCGTAATCAAACAG[G/T]GCCCTGGAGGGCAAGTGGCTATCGG
Phenotype
MIM: 602887
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DLG4 PubMed Links

Gene Details

Gene
DLG4
Gene Name
discs large MAGUK scaffold protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128827.2 1432 Silent Mutation NP_001122299.1
NM_001321074.1 1432 Silent Mutation NP_001308003.1
NM_001321075.1 1432 Silent Mutation NP_001308004.1
NM_001321076.1 1432 Silent Mutation NP_001308005.1
NM_001321077.1 1432 Silent Mutation NP_001308006.1
NM_001365.4 1432 Silent Mutation NP_001356.1
XM_005256491.1 1432 Silent Mutation XP_005256548.1
XM_011523698.1 1432 Silent Mutation XP_011522000.1
XM_011523699.2 1432 Silent Mutation XP_011522001.1
XM_011523702.1 1432 Silent Mutation XP_011522004.1
XM_017024288.1 1432 Silent Mutation XP_016879777.1
XM_017024289.1 1432 Silent Mutation XP_016879778.1
XM_017024290.1 1432 Silent Mutation XP_016879779.1

View Full Product Details