Product Details

SNP ID
rs201054615
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40389634 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGTCCATATGGAAGTCATCACTC[A/T]CCCCCTTGGATTTCTAAAAGAGAAA
Phenotype
MIM: 126430
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TOP2A PubMed Links

Gene Details

Gene
TOP2A
Gene Name
topoisomerase (DNA) II alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001067.3 4582 Missense Mutation GAG,GTG E1494V NP_001058.2
XM_005257632.1 4582 Missense Mutation GAG,GTG E1482V XP_005257689.1
XM_011525165.1 4582 Intron XP_011523467.1

View Full Product Details