Product Details

SNP ID
rs201634767
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28703719 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCTGGACAGTTCTCCCTGCCCCC[A/G]GCCATTGTAACTGTCCTCGCTCTCC
Phenotype
MIM: 601333
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PROCA1 PubMed Links

Gene Details

Gene
PROCA1
Gene Name
protein interacting with cyclin A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304949.1 1064 Missense Mutation NP_001291878.1
NM_001304951.1 1064 Missense Mutation NP_001291880.1
NM_001304952.1 1064 Missense Mutation NP_001291881.1
NM_001304953.1 1064 Missense Mutation NP_001291882.1
NM_001304954.1 1064 Missense Mutation NP_001291883.1
NM_152465.2 1064 Missense Mutation NP_689678.1
XM_005257927.4 1064 Missense Mutation XP_005257984.1
XM_006721723.3 1064 Missense Mutation XP_006721786.1
XM_011524398.2 1064 Missense Mutation XP_011522700.1
XM_011524399.2 1064 Missense Mutation XP_011522701.1
XM_011524400.2 1064 Missense Mutation XP_011522702.1
XM_011524401.1 1064 Missense Mutation XP_011522703.1
XM_017024237.1 1064 Missense Mutation XP_016879726.1
Gene
SUPT6H
Gene Name
SPT6 homolog, histone chaperone
There are no transcripts associated with this gene.

View Full Product Details