Product Details

SNP ID
rs200676913
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8340313 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCAGGTGTTGGCCTCTGAGCTC[A/T]GCCTGGTTGCCTTTATCCTACTATT
Phenotype
MIM: 610097
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ODF4 PubMed Links

Gene Details

Gene
ODF4
Gene Name
outer dense fiber of sperm tails 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319953.1 444 Intron NP_001306882.1
NM_153007.4 444 Missense Mutation AGC,TGC S88C NP_694552.2
XM_005256476.3 444 Intron XP_005256533.1
XM_011523688.1 444 Intron XP_011521990.1

View Full Product Details