Product Details

SNP ID
rs200884166
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8174610 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCACGGATGCACTACAGTGAGCC[C/G]CAATGGCTTGGGAAGGCACGAGATT
Phenotype
MIM: 616663 MIM: 616183
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SNORD118 PubMed Links

Gene Details

Gene
SNORD118
Gene Name
small nucleolar RNA, C/D box 118
There are no transcripts associated with this gene.

Gene
TMEM107
Gene Name
transmembrane protein 107
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032354.3 373 Missense Mutation GCG,GGG A94G NP_115730.2
NM_183065.2 373 Missense Mutation GCG,GGG A88G NP_898888.1

View Full Product Details