Product Details

SNP ID
rs202086475
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:32341240 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGAGGGAGTTTCTGCATTTGTG[C/T]CACTTGGGCTTCCGTCCTCACTGTC
Phenotype
MIM: 603428
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C17orf75 PubMed Links

Gene Details

Gene
C17orf75
Gene Name
chromosome 17 open reading frame 75
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022344.3 273 Missense Mutation NP_071739.2
XM_005258022.3 273 Missense Mutation XP_005258079.1
XM_017024940.1 273 UTR 5 XP_016880429.1
XM_017024941.1 273 UTR 5 XP_016880430.1
Gene
MIR632
Gene Name
microRNA 632
There are no transcripts associated with this gene.

Gene
ZNF207
Gene Name
zinc finger protein 207
There are no transcripts associated with this gene.

View Full Product Details