Product Details

SNP ID
rs202223973
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41611152 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTCACGCATCTCATTCAGGATGC[A/G]GCTCAGGTCCACGCCAGGTGCAGCA
Phenotype
MIM: 148067 MIM: 148069
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRT16 PubMed Links

Gene Details

Gene
KRT16
Gene Name
keratin 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005557.3 Intron NP_005548.2
Gene
KRT17
Gene Name
keratin 17
There are no transcripts associated with this gene.

View Full Product Details