Product Details

SNP ID
rs200955863
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:34287153 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAATAGGAAGATACCCCTTCAGC[A/G]ACTAGAGAGCTACAGGAGAATCACC
Phenotype
MIM: 601156
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCL11 PubMed Links

Gene Details

Gene
CCL11
Gene Name
C-C motif chemokine ligand 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002986.2 275 Missense Mutation CAA,CGA Q45R NP_002977.1

View Full Product Details