Product Details

SNP ID
rs201330894
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28906402 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCGGCCCCACTGCCCCCAATCAA[A/G]CTCGAGCTGCTGGCTTTGCAATTGC
Phenotype
MIM: 616157 MIM: 131560
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
DHRS13 PubMed Links

Gene Details

Gene
DHRS13
Gene Name
dehydrogenase/reductase 13
There are no transcripts associated with this gene.

Gene
FLOT2
Gene Name
flotillin 2
There are no transcripts associated with this gene.

Gene
PHF12
Gene Name
PHD finger protein 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033561.1 2635 Silent Mutation AGC,AGT S932S NP_001028733.1
NM_001290131.1 2635 Intron NP_001277060.1
NM_020889.2 2635 Intron NP_065940.1
XM_005258015.3 2635 Silent Mutation AGC,AGT S842S XP_005258072.1
XM_005258016.3 2635 Silent Mutation AGC,AGT S715S XP_005258073.1
XM_011525078.2 2635 Silent Mutation AGC,AGT S910S XP_011523380.1
XM_011525079.2 2635 Silent Mutation AGC,AGT S899S XP_011523381.1
XM_011525082.2 2635 Intron XP_011523384.1
XM_011525083.2 2635 Intron XP_011523385.1
XM_017024903.1 2635 Silent Mutation AGC,AGT S950S XP_016880392.1
XM_017024904.1 2635 Silent Mutation AGC,AGT S901S XP_016880393.1

View Full Product Details