Product Details

SNP ID
rs201365950
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1468028 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCAATCCTCTGCTTGACTTTGGC[A/G]TCCTCCACGATGACGACGGCGTTGG
Phenotype
MIM: 606538
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYO1C PubMed Links

Gene Details

Gene
MYO1C
Gene Name
myosin IC
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080779.1 2860 Silent Mutation NP_001074248.1
NM_001080950.1 2860 Silent Mutation NP_001074419.1
NM_033375.4 2860 Silent Mutation NP_203693.3

View Full Product Details