Product Details

SNP ID
rs200131498
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:24427954 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAAGCAATGGCAGCCATTTATGG[C/T]GAGGAGTGGTGTGTCATTGATGACT
Phenotype
MIM: 615319
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IMPACT PubMed Links

Gene Details

Gene
IMPACT
Gene Name
impact RWD domain protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018439.3 184 Silent Mutation GGC,GGT G24G NP_060909.1
XM_017025832.1 184 Silent Mutation GGC,GGT G6G XP_016881321.1

View Full Product Details