Product Details

SNP ID
rs199715804
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:5891709 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACTGGAGTTGACACCCCCTGGAG[A/C]CCTAGGGTGGCTCCTGGACCGGTTT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TMEM200C PubMed Links

Gene Details

Gene
TMEM200C
Gene Name
transmembrane protein 200C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080209.1 396 Missense Mutation GCT,TCT A119S NP_001073678.1
XM_005258130.4 396 Missense Mutation GCT,TCT A119S XP_005258187.1
XM_011525729.2 396 Missense Mutation GCT,TCT A119S XP_011524031.1

View Full Product Details