Product Details
- SNP ID
-
rs199633290
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:2656247 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGCTCGGACTACGCGGGATTTCGC[A/G]CGTGTGTGTGTCAGGTACGCGAAGG
- Phenotype
-
MIM: 614982
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CBX3P2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2430853] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CBX3P2
- Gene Name
- chromobox 3 pseudogene 2
There are no transcripts associated with this gene.
- Gene
- SMCHD1
- Gene Name
- structural maintenance of chromosomes flexible hinge domain containing 1
View Full Product Details