Product Details

SNP ID
rs200521548
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:2666870 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATGATTTTCACTCTTGATTACAG[A/G]TGAAACTGTTAAAGATGGAGTCACC
Phenotype
MIM: 614982
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMCHD1 PubMed Links

Gene Details

Gene
SMCHD1
Gene Name
structural maintenance of chromosomes flexible hinge domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015295.2 452 Missense Mutation GAT,GGT D88G NP_056110.2
XM_011525642.1 452 Missense Mutation GAT,GGT D88G XP_011523944.1
XM_011525643.2 452 Missense Mutation GAT,GGT D88G XP_011523945.1
XM_011525644.2 452 UTR 5 XP_011523946.1
XM_017025684.1 452 UTR 5 XP_016881173.1

View Full Product Details