Product Details

SNP ID
hCV190411027
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:16198060 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCGCCGTCTACGTGCTGGACCTG[A/G]AGGGCAAGGTACTGAGGGCTCCCCA
Phenotype
MIM: 603535 MIM: 614554
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AP1M1 PubMed Links

Gene Details

Gene
AP1M1
Gene Name
adaptor related protein complex 1 mu 1 subunit
There are no transcripts associated with this gene.

Gene
FAM32A
Gene Name
family with sequence similarity 32 member A
There are no transcripts associated with this gene.

View Full Product Details