Product Details
- SNP ID
-
rs202206620
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:49044069 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCATCCTGCCCTCAGTCGGAATCCA[C/T]GGTGAACAGCTGGATGTCCTGTGGA
- Phenotype
-
MIM: 607461
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DYM
PubMed Links
Gene Details
- Gene
- DYM
- Gene Name
- dymeclin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017653.3 |
2170 |
Missense Mutation |
ATG,GTG |
M666V |
NP_060123.3 |
XM_006722488.2 |
2170 |
UTR 3 |
|
|
XP_006722551.1 |
XM_006722490.2 |
2170 |
Intron |
|
|
XP_006722553.1 |
XM_006722491.2 |
2170 |
Intron |
|
|
XP_006722554.1 |
XM_006722492.3 |
2170 |
Intron |
|
|
XP_006722555.1 |
XM_011526036.1 |
2170 |
Missense Mutation |
CAT,CGT |
H705R |
XP_011524338.1 |
XM_011526037.1 |
2170 |
Missense Mutation |
CAT,CGT |
H704R |
XP_011524339.1 |
XM_011526038.1 |
2170 |
Missense Mutation |
CAT,CGT |
H704R |
XP_011524340.1 |
XM_011526039.1 |
2170 |
Intron |
|
|
XP_011524341.1 |
XM_011526040.1 |
2170 |
Missense Mutation |
CAT,CGT |
H649R |
XP_011524342.1 |
XM_011526041.1 |
2170 |
Missense Mutation |
CAT,CGT |
H644R |
XP_011524343.1 |
XM_011526042.1 |
2170 |
Intron |
|
|
XP_011524344.1 |
XM_017025795.1 |
2170 |
Missense Mutation |
CAT,CGT |
H703R |
XP_016881284.1 |
XM_017025796.1 |
2170 |
Missense Mutation |
CAT,CGT |
H645R |
XP_016881285.1 |
XM_017025797.1 |
2170 |
Missense Mutation |
ATG,GTG |
M665V |
XP_016881286.1 |
XM_017025798.1 |
2170 |
Missense Mutation |
ATG,GTG |
M665V |
XP_016881287.1 |
XM_017025799.1 |
2170 |
Missense Mutation |
ATG,GTG |
M605V |
XP_016881288.1 |
XM_017025800.1 |
2170 |
Missense Mutation |
ATG,GTG |
M604V |
XP_016881289.1 |
XM_017025801.1 |
2170 |
Missense Mutation |
ATG,GTG |
M603V |
XP_016881290.1 |
- Gene
- MIR4744
- Gene Name
- microRNA 4744
There are no transcripts associated with this gene.
View Full Product Details