Product Details

SNP ID
rs201424582
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:3067479 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGAGGTTGCAGTGGTCGTCTGAG[A/G]CCAGGGCCTTCTCGTTCTTCAACCA
Phenotype
MIM: 603508
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYOM1 PubMed Links

Gene Details

Gene
MYOM1
Gene Name
myomesin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003803.3 4902 Missense Mutation GCC,GTC A1614V NP_003794.3
NM_019856.1 4902 Missense Mutation GCC,GTC A1518V NP_062830.1
XM_017026062.1 4902 Missense Mutation GCC,GTC A1602V XP_016881551.1

View Full Product Details