Product Details

SNP ID
rs201380734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:62717180 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATCCAAAATGACTACCTCTTCCA[A/G]CTGGGATTTGGGGAGCTGTGGAGGG
Phenotype
MIM: 609396
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PHLPP1 PubMed Links

Gene Details

Gene
PHLPP1
Gene Name
PH domain and leucine rich repeat protein phosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194449.3 1742 Silent Mutation CAA,CAG Q499Q NP_919431.2
XM_011525886.1 1742 Missense Mutation CAA,CAG Q499Q XP_011524188.1

View Full Product Details