Product Details

SNP ID
rs201258274
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:79904080 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGGATGGCCAGGTCCACCAGCA[A/C]CTGCAGCAGGCCGCACACGGAGAAC
Phenotype
MIM: 605696
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KCNG2 PubMed Links

Gene Details

Gene
KCNG2
Gene Name
potassium voltage-gated channel modifier subfamily G member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012283.1 754 Intron NP_036415.1
Gene
PQLC1
Gene Name
PQ loop repeat containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146343.1 754 UTR 3 NP_001139815.1
NM_001146345.1 754 Missense Mutation GTG,TTG V220L NP_001139817.1
NM_025078.4 754 Missense Mutation GTG,TTG V238L NP_079354.2
XM_005266770.2 754 UTR 3 XP_005266827.1
XM_011526188.1 754 Missense Mutation GTG,TTG V120L XP_011524490.1
XM_017025997.1 754 Missense Mutation GTG,TTG V263L XP_016881486.1
XM_017025998.1 754 Missense Mutation GTG,TTG V245L XP_016881487.1
XM_017025999.1 754 Intron XP_016881488.1
XM_017026000.1 754 Intron XP_016881489.1
XM_017026001.1 754 Missense Mutation GTG,TTG V157L XP_016881490.1
XM_017026002.1 754 Intron XP_016881491.1
XM_017026003.1 754 UTR 3 XP_016881492.1
XM_017026004.1 754 UTR 3 XP_016881493.1
XM_017026005.1 754 Missense Mutation GTG,TTG V120L XP_016881494.1

View Full Product Details