Product Details

SNP ID
rs199882635
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55159062 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATAAGGGGTGTCTAGGGGTTGGG[C/T]CAGACTCCAGTTTTGGAGTCTGACT
Phenotype
MIM: 614566 MIM: 191044 MIM: 191041
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAAF3 PubMed Links

Gene Details

Gene
DNAAF3
Gene Name
dynein axonemal assembly factor 3
There are no transcripts associated with this gene.

Gene
TNNI3
Gene Name
troponin I3, cardiac type
There are no transcripts associated with this gene.

Gene
TNNT1
Gene Name
troponin T1, slow skeletal type
There are no transcripts associated with this gene.

View Full Product Details