Product Details

SNP ID
rs199816045
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40224732 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCAGATACCTACGTGCACCTGG[A/C]CCAGCCAGTCTACCACCAGGGCGCG
Phenotype
MIM: 164731 MIM: 600137
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AKT2 PubMed Links

Gene Details

Gene
AKT2
Gene Name
AKT serine/threonine kinase 2
There are no transcripts associated with this gene.

Gene
CNTD2
Gene Name
cyclin N-terminal domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024877.3 396 Missense Mutation GGC,GTC G116V NP_079153.2
XM_006723395.3 396 Missense Mutation GGC,GTC G184V XP_006723458.2
Gene
MAP3K10
Gene Name
mitogen-activated protein kinase kinase kinase 10
There are no transcripts associated with this gene.

Gene
TTC9B
Gene Name
tetratricopeptide repeat domain 9B
There are no transcripts associated with this gene.

View Full Product Details