Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001197122.1 | 1602 | Missense Mutation | CAC,CGC | H436R | NP_001184051.1 |
NM_001197123.1 | 1602 | UTR 3 | NP_001184052.1 | ||
NM_001197124.1 | 1602 | UTR 3 | NP_001184053.1 | ||
NM_001197125.1 | 1602 | UTR 3 | NP_001184054.1 | ||
NM_001197126.1 | 1602 | UTR 3 | NP_001184055.1 | ||
NM_001197127.1 | 1602 | UTR 3 | NP_001184056.1 | ||
NM_001197128.1 | 1602 | UTR 3 | NP_001184057.1 | ||
NM_001571.5 | 1602 | UTR 3 | NP_001562.1 | ||
XM_006723197.1 | 1602 | Missense Mutation | CAC,CGC | H436R | XP_006723260.1 |
XM_006723198.1 | 1602 | Missense Mutation | CAC,CGC | H436R | XP_006723261.1 |
XM_006723200.1 | 1602 | Missense Mutation | CAC,CGC | H401R | XP_006723263.1 |
XM_006723201.1 | 1602 | Missense Mutation | CAC,CGC | H290R | XP_006723264.1 |
XM_006723202.2 | 1602 | Missense Mutation | CAC,CGC | H290R | XP_006723265.1 |
XM_017026766.1 | 1602 | UTR 3 | XP_016882255.1 | ||
XM_017026767.1 | 1602 | UTR 3 | XP_016882256.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_021228.2 | 1602 | Intron | NP_067051.2 | ||
XM_005259122.4 | 1602 | Intron | XP_005259179.1 | ||
XM_011527194.2 | 1602 | Intron | XP_011525496.1 | ||
XM_017027083.1 | 1602 | Intron | XP_016882572.1 |