Product Details

SNP ID
rs200095477
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:34739527 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCTGTATTCTTCCTGTAAGCAGG[C/T]CTTTCTGTAACTAAGCCATATGTGA
Phenotype
MIM: 606741
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF181 PubMed Links

Gene Details

Gene
ZNF181
Gene Name
zinc finger protein 181
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029997.3 366 Silent Mutation GGC,GGT G45G NP_001025168.2
NM_001145665.1 366 Silent Mutation GGC,GGT G44G NP_001139137.1
XM_005258849.2 366 Silent Mutation GGC,GGT G45G XP_005258906.1
XM_005258850.2 366 Silent Mutation GGC,GGT G44G XP_005258907.1
XM_005258851.3 366 Silent Mutation GGC,GGT G45G XP_005258908.1
XM_006723183.3 366 Silent Mutation GGC,GGT G45G XP_006723246.1
XM_006723184.3 366 Silent Mutation GGC,GGT G44G XP_006723247.1
XM_017026739.1 366 Silent Mutation GGC,GGT G44G XP_016882228.1
XM_017026740.1 366 Silent Mutation GGC,GGT G63G XP_016882229.1

View Full Product Details