Product Details

SNP ID
rs200448071
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7519349 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCTAGACCCCAATCCTGAAGATC[C/T]GGACCCGGTTTCTGAAGACCCAGAG
Phenotype
MIM: 605248
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MCOLN1 PubMed Links

Gene Details

Gene
MCOLN1
Gene Name
mucolipin 1
There are no transcripts associated with this gene.

Gene
ZNF358
Gene Name
zinc finger protein 358
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018083.4 168 Missense Mutation CCG,CTG P36L NP_060553.4
XM_005272460.3 168 Missense Mutation CCG,CTG P36L XP_005272517.1
XM_011527695.1 168 Missense Mutation CCG,CTG P36L XP_011525997.1
XM_017026296.1 168 Missense Mutation CCG,CTG P55L XP_016881785.1

View Full Product Details