Product Details

SNP ID
rs199676039
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57720837 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGGCCTTTCTCCAGTGTGAGTTC[G/T]TTGGTGCAGAACAAGTGTGTGTTTC
Phenotype
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
ZNF671 PubMed Links

Gene Details

Gene
ZNF671
Gene Name
zinc finger protein 671
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321375.1 1841 Silent Mutation NP_001308304.1
NM_001321376.1 1841 Silent Mutation NP_001308305.1
NM_024833.2 1841 Silent Mutation NP_079109.2
XM_017027314.1 1841 Nonsense Mutation XP_016882803.1

View Full Product Details