Product Details

SNP ID
rs199818168
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10575018 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCAAAGTGGGCTCGCATCAAGTA[C/T]TCCTTGCCCCCCTGAGGGAACATGG
Phenotype
MIM: 607309 MIM: 600927
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AP1M2 PubMed Links

Gene Details

Gene
AP1M2
Gene Name
adaptor related protein complex 1 mu 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300887.1 1143 Silent Mutation GAA,GAG E355E NP_001287816.1
NM_005498.4 1143 Silent Mutation GAA,GAG E353E NP_005489.2
Gene
CDKN2D
Gene Name
cyclin dependent kinase inhibitor 2D
There are no transcripts associated with this gene.

Gene
KRI1
Gene Name
KRI1 homolog
There are no transcripts associated with this gene.

View Full Product Details