Product Details

SNP ID
rs199788280
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43626179 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGTGTCCGGAGGGCAGCGCCTG[C/G]TTCTGCGCCTCACAGATGATGATAC
Phenotype
MIM: 609744
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CADM4 PubMed Links

Gene Details

Gene
CADM4
Gene Name
cell adhesion molecule 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145296.1 567 Missense Mutation AAC,AAG N203K NP_660339.1
XM_005258620.2 567 Missense Mutation AAC,AAG N203K XP_005258677.1
XM_017026452.1 567 Missense Mutation AAC,AAG N184K XP_016881941.1
Gene
ZNF428
Gene Name
zinc finger protein 428
There are no transcripts associated with this gene.

View Full Product Details