Product Details

SNP ID
rs200104259
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56664187 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGGGTGAAGGCCTTGGCGCACT[A/G]GCCGCACGCGTAGGGCTTCTCGCCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMIM17 PubMed Links

Gene Details

Gene
SMIM17
Gene Name
small integral membrane protein 17
There are no transcripts associated with this gene.

Gene
ZNF835
Gene Name
zinc finger protein 835
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005850.2 1274 Nonsense Mutation CAG,TAG Q338* NP_001005850.2
XM_005259382.2 1274 Nonsense Mutation CAG,TAG Q338* XP_005259439.1
XM_005259383.3 1274 Nonsense Mutation CAG,TAG Q338* XP_005259440.1

View Full Product Details