Product Details

SNP ID
rs200313786
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57778781 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCATGGAGGGGAAGATGAGGCAG[C/T]ACCTTCTAAGCAGAGCACGTGTATA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF586 PubMed Links

Gene Details

Gene
ZNF586
Gene Name
zinc finger protein 586
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077426.2 385 Missense Mutation CAC,TAC H23Y NP_001070894.1
NM_001204814.1 385 Missense Mutation GCA,GTA A22V NP_001191743.1
NM_017652.3 385 Missense Mutation GCA,GTA A65V NP_060122.2

View Full Product Details