Product Details

SNP ID
rs199580546
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11576719 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGGGCTGAGGGTGGCTCACCAG[C/T]GCTTGGAGATCTTAGAGTATGCAAT
Phenotype
MIM: 171640 MIM: 612248
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACP5 PubMed Links

Gene Details

Gene
ACP5
Gene Name
acid phosphatase 5, tartrate resistant
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111034.2 789 Missense Mutation CAC,CGC H129R NP_001104504.1
NM_001111035.2 789 Missense Mutation CAC,CGC H129R NP_001104505.1
NM_001111036.2 789 Missense Mutation CAC,CGC H129R NP_001104506.1
NM_001322023.1 789 Missense Mutation CAC,CGC H129R NP_001308952.1
NM_001611.4 789 Missense Mutation CAC,CGC H129R NP_001602.1
XM_005259938.1 789 Missense Mutation CAC,CGC H129R XP_005259995.1
XM_011528069.2 789 Missense Mutation CAC,CGC H129R XP_011526371.1
Gene
ZNF627
Gene Name
zinc finger protein 627
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290083.1 789 Intron NP_001277012.1
NM_001290084.1 789 Intron NP_001277013.1
NM_001290085.1 789 Intron NP_001277014.1
NM_145295.3 789 Intron NP_660338.1
XM_011527780.2 789 Intron XP_011526082.1

View Full Product Details