Product Details

SNP ID
rs200140242
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56089036 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGGGGCGCAGACTGGGGCGGGG[G/A]CAGCTTGGTGGGAGGCCAGCTGGGG
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
ZNF787 PubMed Links

Gene Details

Gene
ZNF787
Gene Name
zinc finger protein 787
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002836.3 348 Missense Mutation CCC,TCC P46S NP_001002836.2
XM_011526445.2 348 Missense Mutation CCC,TCC P46S XP_011524747.1

View Full Product Details