Product Details

SNP ID
rs199512530
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11576264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACCTGCAGATTGTGATCGTGGCC[A/G]CACAGGTAGGCAGTGACCCCGTATG
Phenotype
MIM: 171640 MIM: 612248
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACP5 PubMed Links

Gene Details

Gene
ACP5
Gene Name
acid phosphatase 5, tartrate resistant
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111034.2 1117 Silent Mutation TGC,TGT C238C NP_001104504.1
NM_001111035.2 1117 Silent Mutation TGC,TGT C238C NP_001104505.1
NM_001111036.2 1117 Silent Mutation TGC,TGT C238C NP_001104506.1
NM_001322023.1 1117 Silent Mutation TGC,TGT C238C NP_001308952.1
NM_001611.4 1117 Silent Mutation TGC,TGT C238C NP_001602.1
XM_005259938.1 1117 Silent Mutation TGC,TGT C238C XP_005259995.1
XM_011528069.2 1117 Silent Mutation TGC,TGT C238C XP_011526371.1
Gene
ZNF627
Gene Name
zinc finger protein 627
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290083.1 1117 Intron NP_001277012.1
NM_001290084.1 1117 Intron NP_001277013.1
NM_001290085.1 1117 Intron NP_001277014.1
NM_145295.3 1117 Intron NP_660338.1
XM_011527780.2 1117 Intron XP_011526082.1

View Full Product Details