Product Details

SNP ID
rs200177114
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50512171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCAGAAGGCCGTGACGGTGGCG[C/G]AGCCGGTGACCGCGCCTGGCTCGGC
Phenotype
MIM: 614545 MIM: 615324
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
ASPDH PubMed Links
Additional Information
For this assay, SNP(s) [rs12977172] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ASPDH
Gene Name
aspartate dehydrogenase domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024656.2 835 Missense Mutation TCC,TGC S153C NP_001019827.2
NM_001114598.1 835 Missense Mutation TCC,TGC S258C NP_001108070.1
Gene
EMC10
Gene Name
ER membrane protein complex subunit 10
There are no transcripts associated with this gene.

Gene
JOSD2
Gene Name
Josephin domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270639.1 835 Intron NP_001257568.1
NM_001270640.1 835 Intron NP_001257569.1
NM_001270641.1 835 Intron NP_001257570.1
NM_001270686.1 835 Intron NP_001257615.1
NM_138334.3 835 Intron NP_612207.1
XM_011526434.2 835 Intron XP_011524736.1
Gene
LRRC4B
Gene Name
leucine rich repeat containing 4B
There are no transcripts associated with this gene.

View Full Product Details