Product Details

SNP ID
rs200391100
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41708770 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCCCCTCCCCACAGATGGTGCAT[C/G]CCCTGGCAGAGGCTCCTGCTCACAG
Phenotype
MIM: 114890
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CEACAM5 PubMed Links

Gene Details

Gene
CEACAM5
Gene Name
carcinoembryonic antigen related cell adhesion molecule 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291484.2 155 Silent Mutation ATC,ATG I13M NP_001278413.1
NM_001308398.1 155 Silent Mutation ATC,ATG I13M NP_001295327.1
NM_004363.5 155 Silent Mutation ATC,ATG I13M NP_004354.3
XM_005258413.3 155 Missense Mutation ATC,ATG I13M XP_005258470.2
XM_011526322.1 155 Missense Mutation ATC,ATG I13M XP_011524624.1
XM_017026145.1 155 Intron XP_016881634.1
XM_017026146.1 155 Intron XP_016881635.1

View Full Product Details